Learn More
Tocris Bioscience™ JP-2-196 alkyne
Descripción
JP-2-196 alkyne is a covalent RNF126 chemical handle that binds to several other RING E3 ubiquitin ligases including RNF14, RNF40, RNF219, MID2, and LRSAM1. By facilitating E3 ligase recruitment, JP-2-196 alkyne induces proteasomal degradation of diverse target proteins. It serves as a modular scaffold for the design and synthesis of molecular glue degraders. JP-2-196 alkyne is an alkyne functionalized version of JP-2-196 (Cat. No. 8046).
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1-(4-Methoxyphenyl)-4-[4-(2-propyn-1-yl)-1-piperazinyl]-2-butene-1,4-dione |
| CAS | 2901113-25-3 |
| Cantidad | 50 mg |
| Objetivo | Molecular Glues |
| Fórmula molecular | C18H20N2O3 |
| Pureza | 0.95 |