Learn More
Tocris Bioscience™ JNJ DGAT2-A
GREENER_CHOICE
DGAT2 inhibitor
Marca:  Tocris Bioscience™ 5853/10
Encompass
Código de producto. 15777146
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
JNJ DGAT2-A is a DGAT2 inhibitor (IC50 = 140 nM). Exhibits >70-fold selectivity for DGAT2 over DGAT1 and MGAT2.
Especificaciones
Especificaciones
3-Bromo-4-[2-fluoro-4-[[4-oxo-2-[[2-(pyridin-2-yl)ethyl]amino]-1,3-thiazol-5-(4H)ylidene]methyl]phenoxy]benzonitrile
1962931-71-0