Learn More
Tocris Bioscience™ JHU 37160
Descripción
JHU 37160 is a high affinity and highly potent activator of hM3Dq and hM4Di DREADDs (Ki values are 1.9 nM and 3.6 nM for hM3Dq and hM4Di in vitro, respectively; EC50 values are 18.5 nM and 0.2 nM for hM3Dq and hM4Di in vitro, respectively). Displays approximately 25-fold higher affinity for hM4Di compared to DREADD agonist 21 (Cat. No. 5548). Selectively displaces [3H]clozapine from DREADDs in vivo, but not from other clozapine binding sites. Inhibits locomotor activity in mice expressing hM3Dq and hM4Di in D1-expressing neurons, and increases hM3Dq-stimulated locomotion in rats expressing hM3Dq in TH-expressing neurons. Brain penetrant in mice, rats and non-human primates.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 8-Chloro-11-(4-ethylpiperazin-1-yl)-4-fluoro-5 H-dibenzo[b,e][1,4 ]diazepine |
| CAS | 2369979-68-8 |
| Cantidad | 10 mg |
| Objetivo | DREADD Activators |
| Fórmula molecular | C19H20ClFN4 |
| Pureza | 0.98 |