missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JAB1 Polyclonal Antibody

Descripción
Recommended positive controls: HeLa, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (99%), Xenopus laevis (99%), Pig (100%), Chicken (99%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the 'Signalosome', which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
Especificaciones
Especificaciones
| Antígeno | JAB1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.35 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| génica | Cops5 |
| N.º de referencia del gen | O35864, Q92905 |
| Alias de gen | 38 kDa Mov34 homolog; AI303502; COP9 (constitutive photomorphogenic) homolog, subunit 5; COP9 (constitutive photomorphogenic) homolog, subunit 5 (Arabidopsis thaliana); COP9 complex S5; COP9 constitutive photomorphogenic homolog subunit 5; COP9 signalosome complex subunit 5; COP9 signalosome subunit 5; Cops5; CSN5; JAB1; Jun activation domain-binding protein 1; Jun coactivator; Kic2; Kip1 C-terminus-interacting protein 2; MGC3149; Mov34; MOV-34; SGN5; signalosome subunit 5; testis secretory sperm-binding protein Li 231m |
| Símbolos de los genes | Cops5 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?