Learn More
Tocris Bioscience™ J 2156
Descripción
J 2156 is a high affinity human somatostatin receptor 4 (sst4) agonist (Ki = 1.2 nM). Displays >400-fold selectivity for human sst4 over other human sst receptor subtypes. Exhibits 2 to 3-fold more potent agonism in vitro than endogenous ligands somatostatin-14 (Cat. No. 1157) and somatostatin-28 (Cat. No. 1165). Reduces inflammatory airway hyperresponsiveness in acute and chronic airway inflammation models in mice. Exhibits anxiolytic effects in elevated plus maze test in mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (1'S,2S)-4-Amino-N-(1'-carbamoyl-2'-phenylethyl)-2-(4"-methyl-1"-naphthalenesulfonylamino)butanamide trifluoroacetate |
| CAS | 2387505-73-7 |
| Cantidad | 10 mg |
| Fórmula molecular | C24H28N4O4S.C2HF3O2 |
| Pureza | 98% |