Learn More
Tocris Bioscience™ ITE
Descripción
Endogenous aryl hydrocarbon receptor (AhR) agonist (Ki = 3 nM). Decreases Oct4 levels in U87 glioblastoma cells. Induces stem-like cancer cell differentiation in U87 tumor spheres and inhibits ovarian cancer cell proliferation in vitro. Suppresses tumor growth in U87 and OVCAR-3 cell xenografts in mice. Also inhibits TGF-β-induced human myofibroblast differentiation.
Especificaciones
Especificaciones
| Nombre del producto químico o material | ITE |
| CAS | 448906-42-1 |
| Cantidad | 10 mg |
| Fórmula molecular | C14H10N2O3S |
| Formula Weight (peso de la fórmula) | 286.3 |
| Pureza | >98% |
| Peso molecular | 286.305 |