Learn More
Tocris Bioscience™ Isoginkgetin
Descripción
Isoginkgetin is a pre-mRNA splicing inhibitor (IC50 ∽ 30 μM); inhibits major and minor spliceosomes. Suppresses transcriptional elongation. Blocks spliceosome-mediated splicing process at the pre-spliceosome/A complex stage. Does not inhibit CDK9. Inhibits tumor cell invasion. Active in vivo. Cell permeable
Especificaciones
Especificaciones
| Nombre del producto químico o material | 8-[5-(5,7-Dihydroxy-4-oxo-4H-1-benzopyran-2-yl)-2-methoxyphenyl]-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one |
| CAS | 548-19-6 |
| Cantidad | 10 mg |
| Fórmula molecular | C32H22O10 |
| Pureza | 98% |