Learn More
Óxido de titanio y hierro(II), +99,8 % (base metálica), Thermo Scientific Chemicals
Código de producto. 11322238
Cambiar vista
Click to view available options
Cantidad:
50 g
250 g
Tamaño de la unidad:
250 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
11322238 250 g 250 g
11312238 50 g 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 11322238 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor 013137.30
Solicitar formato a granel o personalizado
CAS: 12022-71-8
Iron(II) titanium oxide is used as a chief source of titanium. It is also used to prepare titanium dioxide by reacting sulfuric acid to get titanium oxygen sulfate followed by hydrolysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 12022-71-8
Número MDL MFCD00064704
Sinónimo Iron(II) titanate
Especificaciones
Nombre del producto químico o material Iron(II) titanium oxide
Forma física Polvo de malla -100
Cantidad 250 g
Notas del porcentaje del ensayo (base metálica)
Información de solubilidad Insoluble in water.
Formula Weight (peso de la fórmula) 151.75
Porcentaje de pureza ≥99.8%