Learn More
Ipratropium bromide, 98%, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar J60453.MC
| Cantidad | 100mg |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
| Ipratropium bromide | |
| C20H30BrNO3 | |
| 100mg | |
| ipratropium bromide hydrate, ipratropium bromide monohydrate, sch 1000-br-monohydrate, atrovent tn, atropine isopropyl bromide, unii-j697uz2a9j, 1r,3s,5s,8r-3-3-hydroxy-2-phenylpropanoyl oxy-8-isopropyl-8-methyl-8-azabicyclo 3.2.1 octan-8-ium hydrate bromide, ipratropium bromide usan:usp:inn:ban:jan, ipratropium bromide usan, sch-1000-br monohydrate | |
| [Br-].CC(C)[N+]1(C)[C@H]2CC[C@@H]1CC(C2)OC(=O)C(CO)C1=CC=CC=C1 | |
| 412.37 | |
| 412.37 |
| 22254-24-6 | |
| MFCD00069291 | |
| 14,5073 | |
| LHLMOSXCXGLMMN-CLTUNHJMSA-M | |
| (1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-methyl-8-(propan-2-yl)-8-azabicyclo[3.2.1]octan-8-ium bromide | |
| 16738693 | |
| 98% |