Learn More
Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli, no manual
Especificaciones
Especificaciones
| Cepa bacteriana o de levadura | TOP10 |
| Contenido y almacenamiento | Store One Shot E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
| Incluye | Linearized pCR2.1vector, T4 DNA ligase, 10X ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls, One Shot TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Without manual. |
| N.º de reacciones | 20 |
| Tipo de producto | One Shot TOP10 Chemically Competent E. coli |
| Cantidad | 20 reacciones |
| Vector | TA Cloning Vectors |