Learn More
Invitrogen™ Countess™ Automated Cell Counter, with 101 Boxes of Slides
Código de producto. 10389053
Cambiar vista
Click to view available options
Tipo de producto:
Contador de células automatizado, 1 caja de portaobjetos
Contador de células automatizado, 101 cajas de portaobjetos
Contador de células automatizado, 11 cajas de portaobjetos
Tamaño de la unidad:
cada uno
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tipo de producto unitSize
10389053 Contador de células automatizado, 1 caja de portaobjetos cada uno
10185154 Contador de células automatizado, 11 cajas de portaobjetos cada uno
10497698 Contador de células automatizado, 101 cajas de portaobjetos cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 10389053 Proveedor Invitrogen™ N.º de proveedor C10227
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Includes
  • One Countess Automated Cell Counter, 101 boxes of 50 slides each (1050 total slides), 2 x 100mL bottles of Trypan blue stain, Power cord with 4 adaptors and Quick Reference Guide (C10311)
  • One Countess Automated Cell Counter, one box of 50 slides (100 total slides), 2 x 1mL vials of Trypan blue stain, power cord with 4 adaptors, and Quick Reference Guide (C10227)
  • One Countess Automated Cell Counter, 11 boxes of 50 slides each (550 total slides), 22 x 1mL vials of Trypan blue stain, Power cord with 4 adaptor and Quick Reference Guide (C10310)
Benchtop instrument for easy cell and viability counts in as little as 30 seconds
  • For verifying Number of Cells, Live Cells/mL, Dead Cells/mL, Cell Size
  • Compatible with Countess cell counting chamber slides
  • Dimensions: D x W x H: 20 x 27 x 19cm (7.8 x 10.6 x 7.4 in.)
Cell Counting, Viability and Cryopreservation
TRUSTED_SUSTAINABILITY
Especificaciones
Dimensiones (Pr x An x Al) 20 x 27 x 19cm
Tensión 100-240 VAC
Frecuencia 50/60 Hz
Tipo de producto Contador de células automatizado, 1 caja de portaobjetos
Incluye One Countess Automated Cell Counter, one box of 50 slides (100 total slides), 2 x 1mL vials of Trypan blue stain, power cord with 4 adaptors, and Quick Reference Guide