missing translation for 'onlineSavingsMsg'
Learn More

INSL3 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7208564 Shop All Proteintech Products
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Unit Size:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16838024 20 μL 20 microlitros
16828024 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16838024 Supplier Proteintech Supplier No. 179971AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Seems to play a role in testicular function. May be a trophic hormone with a role in testicular descent in fetal life. Is a ligand for LGR8 receptor.

Specifications

Antígeno INSL3
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.26 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica INSL3
N.º de referencia del gen P51460
Alias de gen INSL3, Insulin like 3, insulin like 3 (Leydig cell), Ley I L, Leydig insulin like peptide, Relaxin like factor, RLF, RLNL
Símbolos de los genes INSL3
Especie del huésped Rabbit
Inmunógeno INSL3 Fusion Protein Ag12455
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3640
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.