missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A H5N1 HA Antibody Cocktail

Descripción
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.
Especificaciones
Especificaciones
| Antígeno | Influenza A H5N1 HA |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Cocktail |
| Clon | 1D10 |
| Concentración | 1.04 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| Alias de gen | flu hemagglutinin; influenza hemagglutinin |
| Especie del huésped | Mouse |
| Inmunógeno | Influenza A virus H5N1 HA (Hemagglutinin) (A/Hatay/2004/(H5N1)). The exact sequence is proprietary. |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?