Learn More
STEMCELL Technologies (-)-Indolactam V
Descripción
- (-)-Indolactam V is an indole alkaloid compound that activates protein kinase C (PKC)
- It binds to the α, β, γ, δ, ε, and η isozymes of PKC with Ki values of 11, 6, 19, 8, 22, and 16 nM respectively (Kazanietz et al., Masuda et al.)
- Promotes differentiation to human and mouse pancreatic precursors from pluripotent stem cell-derived definitive endoderm (Borowiak et al., Chen et al., Thatava et al.)
Especificaciones
Especificaciones
| CAS | 90365-57-4 |
| Cantidad | 1 mg |
| Fórmula molecular | C17H23N3O2 |
| Pureza | ≥97% |