Learn More
Tocris Bioscience™ Indisulam
Descripción
Indisulam is an acts as a molecular glue to induce proteosomal degradation of mRNA splicing factor RBM39 (also designated CAPERα, HCC1, FSAP59, and RNPC2), via binding to DCAF15. Acts as a pre-mRNA splicing modulator (SPLAMs; splicing inhibitor sulfonamides), causes aberrant pre-mRNA splicing. Suppresses proliferation of cancer cell lines. Reduces viability of HCT-116 cells (IC50 = 0.56 μM). Induces cell cycle arrest in the G1 phase in cancer cell lines. Also a high affinity carbonic anhydrase isozyme XII (hCA XII) inhibitor (Ki = 3.0-5.7 nM).
Especificaciones
Especificaciones
| Nombre del producto químico o material | N1-(3-Chloro-1 H-indol-7-yl)-1,4-benzenedisulfonamide |
| CAS | 165668-41-7 |
| Cantidad | 25 mg |
| Objetivo | Ubiquitin Ligase (E3) Inhibitors |
| Fórmula molecular | C14H12ClN3O4S2 |
| Formula Weight (peso de la fórmula) | 385.84 |
| Tipo de producto | pre-mRNA Splicing Modulator |
| Pureza | 98% |