Learn More
Impurity of Doxercalciferol, MedChemExpress
Código de producto. 16488218
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
25 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16488218 10 mg 10 miligramos
16423776 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16488218 Proveedor Medchem Express N.º de proveedor HY76937
MedChemExpress Impurity of Doxercalciferol is an impurity of doxercalciferol, which is a synthetic analog of ergocalciferol (vitamin D2), used as a drug for secondary hyperparathyroidism and metabolic bone disease, and it suppresses parathyroid synthesis and secretion.
Impurity of Doxercalciferol is an impurity of doxercalciferol, which is a synthetic analog of ergocalciferol (vitamin D2), used as a drug for secondary hyperparathyroidism and metabolic bone disease, and it suppresses parathyroid synthesis and secretion.
Especificaciones
Nombre del producto químico o material Impurity of Doxercalciferol
CAS 127516-23-8
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C28H44O2
Cantidad 10 mg
Información de solubilidad DMSO : 100 mg/mL (242.34 mM; Need ultrasonic)
SMILES C=C([C@H](O)C[C@H](O)C/1)C1=C/C=C2[C@@]3([H])[C@@](CCC/2)(C)[C@]([C@H](C)/C=C/[C@H](C)C(C)C)([H])CC3
Peso molecular (g/mol) 412.65
Formula Weight (peso de la fórmula) 412.65
Porcentaje de pureza 96.08%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Metabolism-sugar/lipid metabolism