missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
Specifications
Specifications
| Antígeno | IL2RG |
| Aplicaciones | Flow Cytometry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.14 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | IL2RG |
| N.º de referencia del gen | P31785, P34902 |
| Alias de gen | CD132, IL 2 receptor subunit gamma, IL 2R subunit gamma, IL 2RG, IL2RG, IMD4, p64, SCIDX, SCIDX1 |
| Símbolos de los genes | IL2RG |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?