Learn More
PeproTech IL-2 Polyclonal Antibody, PeproTech®, Invitrogen™
Descripción
AA Sequence of recombinant protein: PTSSSTSSST AEAQQQQQQQ QQQQQHLEQL LMDLQELLSR MENYRNLKLP RMLTFKFYLP KQATELKDLQ CLEDELGPLR HVLDLTQSKS FQLEDAENFI SNIRVTVVKL KGSDNTFECQ FDDESATVVD FLRRWIAFCQ SIISTSPQ. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-2. Anti-Murine IL-2-specific antibody was purified by affinity chromatography employing an immobilized Murine IL-2 matrix. Sandwich ELISA: To detect Murine IL-2 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine IL-2 (500-P111Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine IL-2. Western Blot: To detect Murine IL-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine IL-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Especificaciones
Especificaciones
| Antígeno | IL-2 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1-1.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | IL2 |
| N.º de referencia del gen | P04351 |
| Alias de gen | aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF |
| Símbolos de los genes | IL2 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.