Learn More
IKZF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12016-1-AP-150UL
 View more versions of this product
Product Code. 16849473
  • 410.00€ / 150 microlitros
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
Specifications
Specifications
IKZF1
Polyclonal
Unconjugated
IKZF1
DNA binding protein Ikaros, hIk 1, Hs.54452, IK1, IKAROS, IKZF1, LYF1, PRO0758, ZNFN1A1
Rabbit
Antigen Affinity Chromatography
RUO
10320, 22778, 305501
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q03267, Q13422
IKZF1
IKZF1 Fusion Protein Ag2631
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.