Learn More
Iguratimod, MedChemExpress
Código de producto. 16473277
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16473277 50 mg 50 miligramos
16429787 5 mg 5 miligramos
16417268 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16473277 Proveedor Medchem Express N.º de proveedor HY17009
MedChemExpress Iguratimod is an antirheumatic agent, acts as an inhibitor of COX-2, with an IC50 of 20 μM (7.7 μg/mL), but shows no effect on COX-1. Iguratimod also inhibits macrophage migration inhibitory factor (MIF) with an IC50 of 6.81 μM.
Iguratimod is an antirheumatic agent, acts as an inhibitor of COX-2, with an IC50 of 20 μM (7.7 μg/mL), but shows no effect on COX-1. Iguratimod also inhibits macrophage migration inhibitory factor (MIF) with an IC50 of 6.81 μM.
Especificaciones
Nombre del producto químico o material Iguratimod
CAS 123663-49-0
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C17H14N2O6S
Cantidad 50 mg
Sinónimo T614
Información de solubilidad DMSO : 33.33 mg/mL (89.03 mM; Need ultrasonic)
SMILES O=C1C2=CC(OC3=CC=CC=C3)=C(C=C2OC=C1NC([H])=O)NS(=O)(C)=O
Peso molecular (g/mol) 374.37
Formula Weight (peso de la fórmula) 374.37
Porcentaje de pureza 98.35%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation