Learn More
Honeywell Safety Products™ Ignite Glasses
Código de producto. 12320160
Cambiar vista
Click to view available options
Color de marco:
Negro, plata
Rojo/plata
Tamaño de la unidad:
cada uno
paquete de 10
Tinción de lente:
Gris
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Color de marco unitSize Tinción de lente
12320160 Rojo/plata paquete de 10 -
12970345 Negro, plata cada uno -
11847070 Negro, plata paquete de 10 -
12607363 Rojo/plata cada uno -
12617363 Rojo/plata cada uno Gris
12647363 Negro, plata cada uno Gris
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Código de producto. 12320160 Proveedor Honeywell Safety Products™ N.º de proveedor 1017081
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Lightweight, frameless, distortion free, unobstructed vision safety glasses. Honeywell Safety Products™ Ignite Safety Glasses offer a comfortable and secure fit.
  • Nine-base, wraparound lens design.
  • Close-to-the-face protection and superior coverage.
  • Lightweight frameless design.
  • All-day comfort and180° of distortion free, unobstructed vision.
  • Adjustable nose-piece.
  • Reduced slippage and personalized, comfortable, secure fit.
TRUSTED_SUSTAINABILITY
Especificaciones
Certificaciones/conformidad EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1775, EC Attestation Number EC1775
Revestimiento de lente Anti-fog, Impact-resistant
Tipo de producto Gafas de seguridad
Para utilizar con (aplicación) Indoor use. Automotive, Building, Construction, Catering, Food, Industrial Cleaning, Maintenance, Service, Ship Building, Textile, Transportation and Utility industries.
Color de marco Rojo/plata
N.° por paquete 1 Ea.
Lens Material Policarbonato
Material del bastidor Polyamide