Learn More
Tocris Bioscience™ (+)-Igmesine hydrochloride
Descripción
(+)-Igmesine hydrochloride is a selective σ1 receptor ligand (KD = 19.1 nM), with little or no activity at σ2 receptors (IC50 > 1000 nM). Inhibits the NMDA-induced increase in cGMP in a concentration-dependent manner (IC50 value is approximately 100 nM). Weak inhibitor of brain 5-HT uptake in vitro. Exhibits neuroprotective and antidepressant effects.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (R)-(+)-N-Cyclopropylmethyl-α-ethyl-N-methyl-α-[(2E)-3-phenyl-2-propenyl)benzenemethanamine hydrochloride |
| CAS | 130152-35-1 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 10 mg |
| Solubilidad | Soluble to 50mM in DMSO |
| Fórmula molecular | C23H29N.HCl |
| Formula Weight (peso de la fórmula) | Observed MW: 355.94 |
Para uso exclusivo en investigación