missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bethyl Laboratories IDO1 Recombinant Rabbit Monoclonal Antibody (BLR040F)
Descripción
IDO1 Recombinant Monoclonal Antibody for IP, Western Blot
IDO1 is an intracellular heme-containing enzyme that catalyzes the oxidative cleavage of the indole ring of several important regulatory molecules like tryptophan, serotonin, and melatonin. By doing this, IDO1 initiates the production of biologically active metabolites, commonly referred to as kynurenines. IDO1 is widely expressed in a variety of human tissues as well as in macrophages and dendritic cells (DCs). In inflammation, interferons (IFNs) act on specific receptors to trigger IDO1 induction. The production of IFN-gamma and induction of IDO1 represent important antimicrobial mechanisms. Degradation and depletion of tryptophan by IDO1 inhibits the growth of viruses, bacteria and parasites. Furthermore, IDO1 plays a complex and crucial role in immunoregulation during infection, pregnancy, autoimmunity, transplantation, and neoplasia.
Especificaciones
Especificaciones
| Antígeno | IDO1 |
| Aplicaciones | Immunoprecipitation, Western Blot |
| Clasificación | Recombinant Monoclonal |
| Clon | BLR040F |
| Concentración | 400 μg/mL |
| Conjugado | Unconjugated |
| Formulación | BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8 to 8.6 |
| génica | Ido1 |
| N.º de referencia del gen | P14902 |
| Alias de gen | EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?