Learn More
Tocris Bioscience™ Ibrutinib
Descripción
Ibrutinib is a potent and selective BTK inhibitor (IC50= 0.5 nM). Selective for BTK against a screening panel of kinase enzymes. Inhibits autophosphorylation of BTK, phosphorylation of PLCγ, and phosphorylation of ERK (IC50 of 11 nM, 29 nM and 13 nM, respectively). Also inhibits ErbB4 (IC50 = 0.25-3.4 nM). Induces cytotoxicity of chronic lymphocytic leukemia (CLL) cells. Reduces clinical arthritis scores in a mouse model.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-2-propen-1-one |
| CAS | 936563-96-1 |
| Sinónimo | Agammaglobulinaemia tyrosine kinase, AGMX1, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, tyrosine-protein kinase |
| Cantidad | 10 mg |
| Fórmula molecular | C25H24N6O2 |
| Formula Weight (peso de la fórmula) | 440.5 |
| Pureza | 98% |