Learn More
I-BRD9, MedChemExpress
Código de producto. 16487566
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16487566 100 mg 100 miligramos
16467668 10 mg 10 miligramos
16463677 50 mg 50 miligramos
16463626 25 mg 25 miligramos
16420138 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16487566 Proveedor Medchem Express N.º de proveedor HY18975
MedChemExpress I-BRD9 is a selective cellular chemical probe of bromodomain-containing protein 9 (BRD9) with pIC50 value of 7.3 μM. I-BRD9 has high selectivity for bromodomain and extra terminal domain (BET) family and highly homologous bromodomain-containing protein 7 (BRD7). I-BRD9 can be used to identify genes regulated by BRD9 in Kasumi-1 cells involved in oncology and immune response pathways.
I-BRD9 is a selective cellular chemical probe of bromodomain-containing protein 9 (BRD9) with pIC50 value of 7.3 μM. I-BRD9 has high selectivity for bromodomain and extra terminal domain (BET) family and highly homologous bromodomain-containing protein 7 (BRD7). I-BRD9 can be used to identify genes regulated by BRD9 in Kasumi-1 cells involved in oncology and immune response pathways.
Especificaciones
Nombre del producto químico o material I-BRD9
Forma física Solid
CAS 1714146-59-4
Color Off-White
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C22H22F3N3O3S2
Para utilizar con (aplicación) Cancer-programmed cell death
Información de solubilidad DMSO : 13.89 mg/mL (27.92 mM; Need ultrasonic)
SMILES N=C(C(S1)=CC2=C1C(C3=CC=CC(C(F)(F)F)=C3)=CN(CC)C2=O)NC(CC4)CCS4(=O)=O
Peso molecular (g/mol) 497.55
Formula Weight (peso de la fórmula) 497.55
Porcentaje de pureza 95.0%