Learn More
Tocris Bioscience™ I-BET 762
Código de producto. 18094322
Cambiar vista
Click to view available options
Cantidad:
5 mg
25 mg
Tamaño de la unidad:
25 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18094322 25 mg 25 miligramos
18054232 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18094322 Proveedor Tocris Bioscience™ N.º de proveedor 6521/25
 Artículo restringido
Fecha estimada de envoi: 11-08-2026
Añadir a la cesta
Añadir a la cesta
Potent and high affinity BET bromodomain inhibitor; anti-inflammatory; orally bioavailable.
I-BET 762 is a potent and high affinity BET bromodomain inhibitor (IC50 = 32.5 - 42.5 nM; Kd = 50.5 - 61.3 nM). Inhibits myeloma cell proliferation. Attenuates transcription of oncogenic MYC and suppresses key inflammatory genes. Exhibits efficacy in a range of oncology and immunoinflammatory models. Displays antiproliferative effect in vitro and in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY
Especificaciones
Nombre del producto químico o material (4S)-6-(4-Chlorophenyl)-N-ethyl-8-methoxy-1-methyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine-4-acetamide
CAS 1260907-17-2
Cantidad 25 mg
Objetivo Bromodomain Inhibitors
Fórmula molecular C22H22ClN5O2
Pureza 0.98