Learn More
Hidroxibupropión-d6, TRC
Product Code. 30408322
Change view
Click to view available options
Cantidad:
1 mg
Unit Size:
1 miligramo
Product Code. Cantidad unitSize
30408322 1 mg 1 miligramo
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30408322 Supplier Toronto Research Chemicals Supplier No. TRCH8306771MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Hydroxy Bupropion-d6
CAS 1216893-18-3
Notas de grado de pureza HPLC
Fórmula molecular C13H12D6ClNO2
Cantidad 1 mg
Sinónimo 2-Hydroxy-2-(3’-chlorophenyl)-3,5,5-trimethylmorpholine-d6
Fórmula InChI InChI=1S/C13H18ClNO2/c1-9-13(16,17-8-12(2,3)15-9)10-5-4-6-11(14)7-10/h4-7,9,15-16H,8H2,1-3H3/i2D3,3D3
SMILES OC1(C2=CC(Cl)=CC=C2)C(C)NC(C([2H])([2H])[2H])(C([2H])([2H])[2H])CO1
Peso molecular (g/mol) 261.78
Formula Weight (peso de la fórmula) 261.1403
Porcentaje de pureza >95