Learn More
Hydrocortisone buteprate, MedChemExpress
Código de producto. 16494088
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16494088 10 mg 10 miligramos
16470447 50 mg 50 miligramos
16405066 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16494088 Proveedor Medchem Express N.º de proveedor HY106673
MedChemExpress Hydrocortisone buteprate (Hydrocortisone probutate) is a medium potent, non-halogenated double-ester of hydrocortisone with a favorable benefit/risk ratio for the treatment of inflammatory skin disorders. Hydrocortisone buteprate (Hydrocortisone probutate) is available as a 0.1% cream or ointment formulation.
Hydrocortisone buteprate (Hydrocortisone probutate) is a medium potent, non-halogenated double-ester of hydrocortisone with a favorable benefit/risk ratio for the treatment of inflammatory skin disorders. Hydrocortisone buteprate (Hydrocortisone probutate) is available as a 0.1% cream or ointment formulation.
Especificaciones
Nombre del producto químico o material Hydrocortisone buteprate
CAS 72590-77-3
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C28H40O7
Cantidad 10 mg
Sinónimo Hydrocortisone probutate HBP
Información de solubilidad DMSO : 250 mg/mL (511.66 mM; Need ultrasonic)
SMILES CCC(OCC([C@@]1([C@@]2([C@@]([C@@]3([H])[C@]([C@@]4(C(CC3)=CC(CC4)=O)C)([H])[C@@H](O)C2)([H])CC1)C)OC(CCC)=O)=O)=O
Peso molecular (g/mol) 488.61
Formula Weight (peso de la fórmula) 488.61
Porcentaje de pureza 98.11%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation