Learn More
Human PTGER1 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18741527
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18741527 25 μg 25 microgramos
18771347 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18741527 Proveedor R&D Systems N.º de proveedor MAB10008SP
Mouse Monoclonal Antibody
PTGER1 Monoclonal specifically detects PTGER1 in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PTGER1
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
Clon 998412
Conjugado Unconjugated
Dilución Immunohistochemistry 5-25 μg;/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen P34995
Alias de gen EP1, PGE receptor EP1 subtype, PGE receptor, EP1 subtype, PGE2 receptor EP1 subtype, prostaglandin E receptor 1 (subtype EP1), 42kD, prostaglandin E receptor 1 (subtype EP1), 42kDa, prostaglandin E receptor 1, subtype EP1, prostaglandin E2 receptor EP1 subtype, Prostanoid EP1 receptor
Especie del huésped Mouse
Inmunógeno Synthetic peptide containing human PTGER1, Accession No. P34995
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Primario o secundario Primary
ID de gen (Entrez) 5731.0
Especificidad de la prueba Detects human PTGER1 in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Formulario Lyophilized
Isotype IgG1