Learn More
R&D Systems™ Human PCYT1A (NP_005008.2) VersaClone cDNA
Especificaciones
Especificaciones
| génica | PCYT1A |
| Número de acceso | NP_005008.2 |
| Concentración | 10 μg at 0.2 μg/μL |
| Formulación | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Tipo de inserto | 1117 bp |
| ID de gen (Entrez) | 5130 |
| Alias de gen | CCT A, CCT-alpha, choline-phosphate cytidylyltransferase A, CT, CT A, CTA, CTP:phosphocholine cytidylyltransferase A, CTPCT, CTPCTphosphate cytidylyltransferase 1, choline, alpha isoform, EC 2.7.7, EC 2.7.7.15, PCYT1, PCYT1CCTA, phosphate cytidylyltransferase 1, choline, alpha, Phosphorylcholine transferase A, SMDCRD |
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Especie | Human |
| Cantidad | 10 μg |