missing translation for 'onlineSavingsMsg'
Learn More

Human Nephrin Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18775657
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
25 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18775657 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18775657 Proveedor R&D Systems N.º de proveedor AF4269SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Sheep Polyclonal Antibody

Nephrin Polyclonal specifically detects Nephrin in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Nephrin
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 1-15 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen O60500
Alias de gen CNF, nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, NPHS1, Renal glomerulus-specific cell adhesion receptor
Símbolos de los genes NPHS1
Especie del huésped Sheep
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human Nephrin Gln23-Thr1029 Accession # O60500
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4868
Especificidad de la prueba Detects human Nephrin in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Nephrin is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.