Learn More
Human/Mouse Cleaved Caspase-3 (Asp175) Antibody, R&D Systems™
Código de producto. 18071862
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
18071862 50 μg 50 microgramos
1 options
Código de producto. 18071862 Proveedor R&D Systems N.º de proveedor MAB835
Rabbit Monoclonal Antibody has been used in 52 publications
Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Caspase-3
Aplicaciones Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Clasificación Monoclonal
Clon 269518
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.5 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen U26943
Alias de gen Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
Símbolos de los genes CASP3
Especie del huésped Rabbit
Inmunógeno KLH-conjugated human Caspase-3 synthetic peptide CRGTELDCGIETD Accession # U26943
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 836
Especificidad de la prueba Detects human and mouse Caspase-3 cleaved at Asp175. No cross-reactivity was detected with the full-length procaspase-3 or other caspases.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human, Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG