Learn More
R&D Systems™ Human mGluR6 (AAB82068) VersaClone cDNA
Descripción
The Human mGluR6 (AAB82068) VersaClone cDNA clone contains the complete ORF for mGluR6, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The mGluR6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Información de órdenes
Ships at ambient temperature.
Especificaciones
Especificaciones
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Cantidad | 10 μg |
For Reasearch Use Only; Not for Use in Humans