Learn More
Invitrogen™ Human IL-2 ELISA Kits
Descripción
Assay IL-2 (human interleukin-2) levels in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples with our Invitrgen™ Human IL-2 ELISA Kits.
Target: Natural and recombinant human interleukin-2 (IL-2)
Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: Absorbance at 450nm minus absorbance at 550nm
Assay Range: 38.4 to 1500pg/mL
Sensitivity: < 6pg/mL
Precision: Intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 4 hours
Sample Size: 50μL
Specificity: No significant cross-reaction with human IL-1α, IL-1β, IL-3, IL-4, IL-6, IL-7, IL-8, TNFα, TNFβ, GM-CSF, INFγ, or mouse IL-2
Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).
Especificaciones
Especificaciones
| Intervalo del ensayo | 38.4 to 1500pg/mL |
| Sensibilidad del ensayo | <6pg/mL |
| Descripción | Human IL-2 ELISA Reagent Kit |
| Formato del kit de inmunoensayo | Colorimetric ELISA (TMB substrate) with Plates |
| Tipo de muestra | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Especificidad | No significant cross-reaction with human IL-1a, IL-1ß, IL-3, IL-4, IL-6, IL-7, IL-8, TNFa, TNFß, GM-CSF, INFg, or mouse IL-2 |
| Suficiente para | 5 x 96 microplate wells |
| Contenido del kit | TMB Substrate Solution, 55mL, Streptavidin-HRP Conjugate, 0.25mL, Stop Solution, 55mL, IL-2 Detection Antibody, 1 vial, Human IL-2 Standard, 5 vials, IL-2 Coating Antibody, 1 vial |
| Medición | Absorbancia a 450 nm menos absorbancia a 550 nm |
| Precisión | CV del intraensayo <10 %; CV del interensayo <10 % |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.