Learn More
Human Endoglycan/PODXL2 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18013352
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
18013352 100 μg 100 microgramos
1 options
Código de producto. 18013352 Proveedor R&D Systems N.º de proveedor AF1524
Goat Polyclonal Antibody
Endoglycan/PODXL2 Polyclonal specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Endoglycan/PODXL2
Aplicaciones Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
N.º de referencia del gen Q9NZ53
Alias de gen endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2, PODXL2
Símbolos de los genes PODXL2
Especie del huésped Goat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human Endoglycan/PODXL2 Gly33-Thr500 Accession # Q9NZ53
Método de purificación Affinity Purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 50512
Especificidad de la prueba Detects human Endoglycan/PODXL2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human PODXL.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG