missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APOB native Protein from human plasma

Código de producto. 16202270
Change view
Click to view available options
Cantidad:
500 μg
Tamaño de la unidad:
500 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16202270 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16202270 Proveedor Abnova™ N.º de proveedor P5328.500ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Used for SDS-PAGE

This gene product is the main apolipoprotein of chylomicrons and low density lipoproteins. It occurs in plasma as two main isoforms, apoB-48 and apoB-100: the former is synthesized exclusively in the gut and the latter in the liver. The intestinal and the hepatic forms of apoB are encoded by a single gene from a single, very long mRNA. The two isoforms share a common N-terminal sequence. The shorter apoB-48 protein is produced after RNA editing of the apoB-100 transcript at residue 2180 (CAA->UAA), resulting in the creation of a stop codon, and early translation termination. Mutations in this gene or its regulatory region cause hypobetalipoproteinemia, normotriglyceridemic hypobetalipoproteinemia, and hypercholesterolemia due to ligand-defective apoB, diseases affecting plasma cholesterol and apoB levels. [provided by RefSeq]

Especificaciones

Para utilizar con (aplicación) SDS-PAGE
Formulación Lyophilized from 1 ml of 10mM sodium deoxycholate, 50mM NaCl, 5 0mM Na
ID de gen (Entrez) 338
Nombre APOB (Human) Native Protein
Método de preparación Purified from low density lipoproteins (LDL) isolated by density gradient ultracentrifugation (1.03 - 1.05 g/mL density range) and by gel filtration chromatography in 10 mM sodium deoxycholate, 50 mM NaCl, 50 mM Na2CO3, pH 10.1.
Método de purificación Density gradient ultracentrifugation and gel filtration chromatography
Cantidad 500 μg
Requisitos de almacenamiento Store at -20°C or lower on dry atmosphere. After reconstitution, store at 2-8°C no longer than 4 weeks. Reconstitution: Dissolve lyophilized Apo B in 100 mL to 1 mL of distilled or deionized water. If dissolving in solvent we recommend using 10 mM sodium deoxycholate, 50 mM NaCl, 50 mM Na2CO3, pH 10 to keep the sodium deoxycholate at an acceptable level instead of water.
Estado normativo RUO
Alias de gen FLDB
Nombre común APOB
Símbolo de gen APOB
Especie Human
Etiqueta de proteína None
Sistema de expresión Purified from low density lipoproteins (LDL) isolated by density gradient ultracentrifugation (1.03 - 1.05 g/mL density range) and by gel filtration chromatography in 10 mM sodium deoxycholate, 50 mM NaCl, 50 mM Na2CO3, pH 10.1.
Formulario Lyophilized
Grado de pureza o calidad ≥95% by SDS-PAGE
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.