Learn More
Lonza™ Human Umbilical Vein Endothelial Cells (HUVEC), Pooled in EGM™−2
Pooled donor cells are ideal for reducing donor variability. Lonza™ Human Umbilical Vein Endothelial Cells (HUVEC), Pooled in EGM™−2 are isolated in EGM™−2 Media with growth factors.
Marca:  Lonza™ C2519A
Código de producto. 11525859
  • 567.00€ / paquete de 1
Descripción
  • Cryopreserved HUVECs shipped as frozen primaries
  • Guaranteed through 15 population doublings
  • Express CD31/105 and von Williebrand Factor VIII
  • Positive for acetylated low density lipoprotein uptake
  • All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots
  • A Certificate of Analysis is provided for each cell lot purchased