Learn More
R&D Systems™ Human ADAM8 (NP_001100.3) VersaClone cDNA
Descripción
Inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Especificaciones
Especificaciones
| génica | ADAM8 |
| Número de acceso | NP_001100.3 |
| Concentración | 10 μg at 0.2 μg/ μL |
| Formulación | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Tipo de inserto | 2488 bp |
| ID de gen (Entrez) | 101 |
| Alias de gen | a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2, MS2disintegrin and metalloproteinase domain-containing protein 8 |
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Especie | Human |
| Cantidad | 10 μg |