missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSP60 Polyclonal Antibody

Descripción
HSP60 Polyclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.
Especificaciones
Especificaciones
| Antígeno | HSP60 |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | whole serum with 5mM sodium azide |
| génica | HSPD1 |
| N.º de referencia del gen | P10809, P63038, P63039 |
| Alias de gen | 60 kDa chaperonin; 60 kDa heat shock protein; 60 kDa heat shock protein, mitochondrial; 60kDa; Chaperonin 60; CPN60; GROEL; heat shock 60kD protein 1 (chaperonin); heat shock 60kDa protein 1; heat shock 60kDa protein 1 (chaperonin); Heat shock protein; heat shock protein 1 (chaperonin); heat shock protein 60; heat shock protein 60 (liver); heat shock protein 65; heat shock protein family D (Hsp60) member 1; heat shock protein, 60 kDa; HLD4; HSP; Hsp60; HSP-60; Hsp60s1; HSP65; HSP-65; Hspd1; Hspd1-30p; HuCHA60; hypothetical protein; I79_011398; mitochondrial matrix protein P1; P1 protein; P60 lymphocyte protein; RCJMB04_7g5; short heat shock protein 60 Hsp60s1; SPG13; Unknown (protein for IMAGE:7943409) |
| Símbolos de los genes | HSPD1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?