missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSH2D Polyclonal Antibody

Código de producto. 16337304
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16337304 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16337304 Proveedor Invitrogen™ N.º de proveedor PA578608

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Positive Control: NCI-H929 Predicted Reactivity: Rhesus Monkey (96%), Chimpanzee (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

T-cell activation requires 2 signals: recognition of antigen by the T-cell receptor and a costimulatory signal provided primarily by CD28 in naive T cells. HSH2 is a target of both of these signaling pathways.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno HSH2D
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
génica HSH2D
N.º de referencia del gen Q96JZ2
Alias de gen Adaptor in lymphocytes of unknown function X; ALX; FLJ14886; hematopoietic SH2 domain containing; hematopoietic SH2 domain-containing protein; Hematopoietic SH2 protein; HSH2; HSH2D
Símbolos de los genes HSH2D
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the center region of human HSH2D. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 84941
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.