Learn More
HQL-79, MedChemExpress
Código de producto. 16466887
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
Tamaño de la unidad:
5 miligramos
16466887 5 mg 5 miligramos
1 options
Código de producto. 16466887 Proveedor Medchem Express N.º de proveedor HY108259
MedChemExpress HQL-79, a potent, selective and orally active human hematopoietic prostaglandin D synthase (H-PGDS) inhibitor, highly selectively inhibits the synthesis of PGD2, and acts as an anti-allergic agent, with a Kd of 0.8 μM and an IC50 of 6 μM. Shows no obvious effect on COX-1, COX-2, m-PGES, or L-PGDS.
HQL-79, a potent, selective and orally active human hematopoietic prostaglandin D synthase (H-PGDS) inhibitor, highly selectively inhibits the synthesis of PGD2, and acts as an anti-allergic agent, with a Kd of 0.8 μM and an IC50 of 6 μM. Shows no obvious effect on COX-1, COX-2, m-PGES, or L-PGDS.
Especificaciones
Nombre del producto químico o material HQL-79
Forma física Solid
CAS 162641-16-9
Color Blanco
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C22H27N5O
Para utilizar con (aplicación) COVID-19-immunoregulation
SMILES N1(CCCC2=NN=NN2)CCC(OC(C3=CC=CC=C3)C4=CC=CC=C4)CC1
Peso molecular (g/mol) 377.48
Formula Weight (peso de la fórmula) 377.48
Porcentaje de pureza 98.0%