missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPGDS inhibitor 2, MedChemExpress
Descripción
HPGDS inhibitor 2 is a highly potent and selective hematopoietic prostaglandin D synthase (H-PGDS) inhibitor with an IC50 of 9.9 nM.
Specifications
Specifications
| Nombre del producto químico o material | HPGDS inhibitor 2 |
| CAS | 2101626-26-8 |
| Color | Off-White |
| Notas de grado de pureza | Research |
| Fórmula molecular | C20H24F2N2O3 |
| Cantidad | 100 mg |
| Información de solubilidad | DMSO : ≥ 150 mg/mL (396.40 mM) |
| SMILES | FC(F)OC1=CC=C2C(N=CC(C(N[C@@H]3CC[C@@H](C(C)(C)O)CC3)=O)=C2)=C1 |
| Peso molecular (g/mol) | 378.41 |
| Formula Weight (peso de la fórmula) | 378.41 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?