missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ hnRNP D Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15971725
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15971725 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15971725 Proveedor Invitrogen™ N.º de proveedor PA575200

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are nucleic acid binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. It localizes to both the nucleus and the cytoplasm. This protein is implicated in the regulation of mRNA stability. Alternative splicing of this gene results in four transcript variants.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno hnRNP D
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
génica HNRNPD
N.º de referencia del gen Q14103, Q60668, Q9JJ54
Alias de gen ARE-binding protein AUFI, type A; Auf1; AUF1A; AU-rich element RNA binding protein 1, 37kDa; AU-rich element RNA-binding factor 1; AU-rich element RNA-binding protein 1; heterogeneous nuclear ribonucleoprotein D; heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa); heterogeneous nuclear ribonucleoprotein D0; hnRNP D0; HNRNPD; hnRNPD0; Hnrpd; P37; RNA binding protein p45AUF1
Símbolos de los genes HNRNPD
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to amino acids 46-98 of Human hnRNP D.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11991, 3184, 79256
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.