missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ hnRNP A2B1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 13434597
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
13434597 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 13434597 Proveedor Invitrogen™ N.º de proveedor PA530061

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A549, HeLa, HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. This gene has been described to generate two alternatively spliced transcript variants which encode different isoforms. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno hnRNP A2B1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.75 mg/mL
Conjugado Unconjugated
Formulación 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
génica HNRNPA2B1
N.º de referencia del gen P22626
Alias de gen 9130414A06Rik; DKFZp779B0244; EMBL:BAF79676.1}; FLJ22720; heterogeneous nuclear ribonucleoprotein A2; heterogeneous nuclear ribonucleoprotein A2/B1; heterogeneous nuclear ribonucleoprotein B0a; heterogeneous nuclear ribonucleoprotein B0b; heterogeneous nuclear ribonucleoprotein B1; heterogeneous nuclear ribonucleoproteins A2/B1; heterogeneous nuclear ribonucleoproteins A2/B1 {ECO:0000312; heterogenous nuclear ribonucleoprotein A2/B1; Hnrnp; hnRNP A2 / hnRNP B1; hnRNP A2/B1; hnrnp-A; HNRNPA2; HNRNPA2B1; hnrnpa2b1 {ECO:0000250; HNRNPB1; HNRPA2; HNRPA2B1; HNRPB1; hypothetical protein; IBMPFD2; nuclear ribonucleoprotein particle A2 protein; OTTHUMP00000202465; OTTHUMP00000202466; RNPA2; SNRPB1; UniProtKB:P22626}
Símbolos de los genes HNRNPA2B1
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the center region of human hnRNP A2B1. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3181
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.