missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ hnRNP A2B1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17875752
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17875752 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17875752 Proveedor Invitrogen™ N.º de proveedor PA5143158

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_002128.1; NP_112533.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

This gene belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. This gene has been described to generate two alternatively spliced transcript variants which encode different isoforms. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno hnRNP A2B1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
génica HNRNPA2B1
N.º de referencia del gen O88569, P22626
Alias de gen 9130414A06Rik; DKFZp779B0244; EMBL:BAF79676.1}; FLJ22720; heterogeneous nuclear ribonucleoprotein A2; heterogeneous nuclear ribonucleoprotein A2/B1; heterogeneous nuclear ribonucleoprotein B0a; heterogeneous nuclear ribonucleoprotein B0b; heterogeneous nuclear ribonucleoprotein B1; heterogeneous nuclear ribonucleoproteins A2/B1; heterogeneous nuclear ribonucleoproteins A2/B1 {ECO:0000312; heterogenous nuclear ribonucleoprotein A2/B1; Hnrnp; hnRNP A2 / hnRNP B1; hnRNP A2/B1; hnrnp-A; HNRNPA2; HNRNPA2B1; hnrnpa2b1 {ECO:0000250; HNRNPB1; HNRPA2; HNRPA2B1; HNRPB1; hypothetical protein; IBMPFD2; nuclear ribonucleoprotein particle A2 protein; OTTHUMP00000202465; OTTHUMP00000202466; RNPA2; SNRPB1; UniProtKB:P22626}
Símbolos de los genes HNRNPA2B1
Especie del huésped Goat
Inmunógeno Peptide with sequence C-DTIEIITDRQS (aa 139-149).
Método de purificación Ammonium sulfate precipitation
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3181, 53379
Especies diana Human, Mouse
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.