missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMBS Monoclonal Antibody (OTI1F1), TrueMAB™, OriGene
Descripción
HMBS Monoclonal antibody specifically detects HMBS in Human samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | HMBS |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI1F1 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | Hmbs |
| N.º de referencia del gen | P08397 |
| Alias de gen | alternative name: porphobilinogen deaminase; HEM3; hemC; HMBS; Hydroxymethylbilane synthase; PBG D; PBGD; PBG-D; PORC; Porphobilinogen deaminase; porphyria, acute; Chester type; pre-uroporphyrinogen synthase; similar to Porphobilinogen deaminase (Hydroxymethylbilane synthase) (HMBS) (Pre-uroporphyrinogen synthase) (PBG-D); T25658; Ups; uroporphyrinogen I synthase; uroporphyrinogen I synthetase; URO-S; Uros1 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?