Learn More
HJ 1189-2021 Mezcla de plaguicidas organofosforados 693 2000 μg/ml en acetona, Dr. Ehrenstorfer
Specifications
Specifications
| Nombre del producto químico o material | Bromophos-methyl∣Chlorpyrifos∣Chlorpyrifos-methyl∣Coumaphos∣Demeton (O+S)∣Diazinon∣Dichlorvos∣Dimethoate∣Ethoprophos∣Fenamiphos∣Fenitrothion∣Fonofos∣Iprobenfos∣Isazofos∣Isocarbofos∣Isofenphos-methyl∣Malathion∣Mevinphos∣Parathion-ethyl∣Parathion-methyl∣Phenthoate∣Phorate∣Phosphamidon∣Profenofos∣Sulfotep∣Terbufos∣Triazophos |
| CAS | 2104-96-3, 2921-88-2, 5598-13-0, 56-72-4, 8065-48-3, 333-41-5 |
| Nombres de analitos o componentes | Bromophos-methyl∣Chlorpyrifos∣Chlorpyrifos-methyl∣Coumaphos∣Demeton (O+S)∣Diazinon∣Dichlorvos∣Dimethoate∣Ethoprophos∣Fenamiphos∣Fenitrothion∣Fonofos∣Iprobenfos∣Isazofos∣Isocarbofos∣Isofenphos-methyl∣Malathion∣Mevinphos∣Parathion-ethyl∣Parathion-methyl∣Phenthoate∣Phorate∣Phosphamidon∣Profenofos∣Sulfotep∣Terbufos∣Triazophos |
| Condiciones de envío | +20°C |
| Certificaciones/conformidad | ISO 17034 |
| Solvent or Matrix | Acetone |
| Cantidad | 1 mL |
| Concentration or Composition (by Analyte or Components) | Bromophos-methyl 2000 μg/mL∣Chlorpyrifos 2000 μg/mL∣Chlorpyrifos-methyl 2000 μg/mL∣Coumaphos 2000 μg/mL∣Demeton (O+S) 2000 μg/mL∣Diazinon 2000 μg/mL∣Dichlorvos 2000 μg/mL∣Dimethoate 2000 μg/mL∣Ethoprophos 2000 μg/mL∣Fenamiphos 2000 μg/mL∣Fenitrothion 2000 μg/mL∣Fonofos 2000 μg/mL∣Iprobenfos 2000 μg/mL∣Isazofos 2000 μg/mL∣Isocarbofos 2000 μg/mL∣Isofenphos-methyl 2000 μg/mL∣Malathion 2000 μg/mL∣Mevinphos 2000 μg/mL∣Parathion-ethyl 2000 μg/mL∣Parathion-methyl 2000 μg/mL∣Phenthoate 2000 μg/mL∣Phorate 2000 μg/mL∣Phosphamidon 2000 μg/mL∣Profenofos 2000 μg/mL∣Sulfotep 2000 μg/mL∣Terbufos 2000 μg/mL∣Triazophos 2000 μg/mL |