Learn More
HDAC2 Mouse anti-Human, Unconjugated, Clone: 1A3E4, Proteintech
Código de producto. 16849645 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16849645 20 μL 20 microlitros
16839645 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16849645 Proveedor Proteintech N.º de proveedor 671651IG20UL
Mouse Monoclonal Antibody
The HDAC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC2. This antibody recognizes human, mouse, rat antigen. The HDAC2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Antígeno HDAC2
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Monoclonal
Clon 1A3E4
Concentración 1000 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 3066
Especies diana Human, Rat, Mouse
Isotype IgG2b