Learn More
Halofuginone, MedChemExpress
Código de producto. 16491648
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16491648 5 mg 5 miligramos
16473926 25 mg 25 miligramos
16439238 10 mg 10 miligramos
16404927 50 mg 50 miligramos
16459166 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16491648 Proveedor Medchem Express N.º de proveedor HYN1584
MedChemExpress Halofuginone (RU-19110), a Febrifugine derivative, is a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18.3 nM.
Halofuginone is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibition of TGF-β activity. Halofuginone is also a potent pulmonary vasodilator by activating Kv channels and blocking voltage-gated, receptor-operated and store-operated Ca2+ channels. Halofuginone has anti-malaria, anti-inflammatory, anti-cancer, anti-fibrosis effects.
Specifications
Nombre del producto químico o material Halofuginone
Forma física Solid
CAS 55837-20-2
Color Blanco
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C16H17BrClN3O3
Para utilizar con (aplicación) COVID-19-anti-virus
Sinónimo RU-19110
Información de solubilidad DMSO : 20 mg/mL (48.23 mM; ultrasonic and adjust pH to 5 with HCl)
SMILES O=C1N(CC(C[C@@H]2NCCC[C@H]2O)=O)C=NC3=C1C=C(Cl)C(Br)=C3
Peso molecular (g/mol) 414.68
Formula Weight (peso de la fórmula) 414.68
Porcentaje de pureza 98.03%