Learn More
MedChemExpress HAEGT
Código de producto. 16402058
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
Tamaño de la unidad:
5 miligramos
16402058 5 mg 5 miligramos
1 options
Código de producto. 16402058 Proveedor MedChemExpress N.º de proveedor HYP1230
MedChemExpress HAEGT is the first N-terminal 1-5 residues of glucagon like peptide-1 (GLP-1) peptide, and the sequence is His-Ala-Glu-Gly-Thr. HAEGT acts as a competitive substrate for probing prime substrate binding sites of human dipeptidyl peptidase-IV (DPP-IV) 1, in which the N-terminal His-Ala is catalyzed cleavage by DPP-IV. HAEGT can be used in the research of diabetes, obesity.
Especificaciones
Pureza 99.26%
CAS 852155-81-8
Contenido y almacenamiento Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Peso molecular 513.5 g/mol
Tipo de producto Peptides